Learn More
Thermo Scientific™ TX-750 Rectangular Bucket Adapters
Codice prodotto. 11804481 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità:
15 mL Conical Tubes
50 mL Conical Tubes
Dimensione della confezione:
pezzi 4
Codice prodotto. 11804481
Marca: Thermo Scientific™ 75003685
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Ordering Information:
Order rectangular buckets (75003614), Thermo Scientific™ ClickSeal™ Biocontainment Lids (75003615) and TX-750 Rotor (75003607) separately. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Tube Adapter
Capacità 50 mL Conical Tubes