Learn More
Thermo Scientific™ TX-750 Rectangular Bucket Adapters
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità:
15 mL Conical Tubes
50 mL Conical Tubes
Dimensione della confezione:
pezzi 4
Descrizione
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Ordering Information:
Order rectangular buckets (75003614), Thermo Scientific™ ClickSeal™ Biocontainment Lids (75003615) and TX-750 Rotor (75003607) separately. Contact your sales representative for more information.
Specifica
Specifica
| Tipo | Tube Adapter |
| Capacità | 15 mL Conical Tubes |