Learn More
Skytec™ TX424 Dual Certified Thin Nitrile Glove
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Dimensione della confezione:
pezzi 100
Descrizione
- High quality nitrile, soft and durable
- High resistance to penetration and projections of many chemicals
- Textured finish on fingertips for improved grip and touch
- Powder free, latex free, non sterile, AQL 1.5
- Chlorinated inside to offer more comfort and reduce friction when donning
- Nitrile has antistatic properties
- Approved for food handling
- Dual labelling: PPE and medical
- Medical device compliant (MDD 93/42/EEC), conforms to EN455 (all parts)
- Tested against chemotherapy drugs according to ASTM D6978-05
- Compliant with ASTM D6319-2019 and ASTM D5151-2019
Specifica
Specifica
| Certificazione/Conformità | CE III , UKCA , EN 374-1:2016 Type B: K,P,T , EN 374-2:2014 Yes , EN 374-5:2016 , EN 421:2010 , EN 455-1:2000 , EN 455-2:2015 Yes , EN 455-3:2015 Yes , EN 455-4:2009 Yes , EN 455 Medical Device , EN 21420:2020 |
| Accelerator Free | No |
| Applicazioni | Healthcare, Industrial, Laboratory |
| AQL | 1.5 |
| Colore | Blue |
| Monouso | Single-use |
| Finitura esterna | Fingertip Textured |
| Finger Thickness (English) | 5.9 mil |
| Finger Thickness (Metric) | 0.15 mm |
| Latex Free | Sì |
| Vedi altri risultati |