Learn More
DuPont™ Tychem™ 2000C Gown
Sfoglia Tutto DuPont Prodotti
Click to view available options
Size:
Medium
2X-Large
Dimensione della confezione:
pezzi 25
Descrizione
- Double cuffs for increased protection and tighter connection with gloves
- Elasticated wrists
- Knitted cuff (inside) for better comfort
- Offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure)
- Used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
- Disposable
Specifica
Specifica
| Size | Medium |
| Colore | Yellow |
| Monouso | Single-use |
| Certificazione/Conformità | (EU) 2016/425, EN 14126, EN 1149-1 |