Learn More
Masterflex™ Tubo da laboratorio Tygon™ R-3603
Click to view available options
Diametro (sistema metrico) interno:
0.8 mm
1.6 mm
2.4 mm
4.8 mm
Diametro (sistema metrico) esterno:
2.4 mm
4.8 mm
5.6 mm
8 mm
Dimensione della confezione:
15m
Pezzo
Descrizione
- Bio-based formulation delivers excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifica
Specifica
| Certificazione/Conformità | Conforme agli standard FDA; conforme ai requisiti USP classe VI e NSF 51, USP Class VI, NSF 51 |
| Diametro (sistema imperiale) interno | 0.09 in. |
| Diametro (sistema metrico) interno | 2.4 mm |
| Materiale | Plastica |
| Diametro (sistema imperiale) esterno | 0.22 in. |
| Diametro (sistema metrico) esterno | 5.6 mm |
| Overall Length (English) | 49.2 ft. |
| Overall Length (Metric) | 15 m |
| Spessore (sistema imperiale) | 0.06 in. |
| Spessore (sistema metrico) | 1,6 mm |
| Vedi altri risultati |