Learn More
DuPont™ Tyvek™ 400 DualCombi Model TD0125S
Codice prodotto. 30305965
Cambia vista
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Dimensione della confezione:
pezzi 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Size unitSize
30305965 2X-Large pezzi 25
30305915 Small pezzi 25
30305939 Medium pezzi 25
30305905 Large pezzi 25
30305898 X-Large pezzi 25
30305962 3X-Large pezzi 25
30305917 4X-Large pezzi 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Codice prodotto. 30305965 Fornitore DuPont™ N. del fornitore D14681416
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Collared coverall with Tyvek at the front and large breathable polypropylene back panel. Stitched external seams in red. Elasticated wrists and ankles.
  • DuPont™ Tyvek™ 400 DualCombi. Collared coverall combining Tyvek™ with a spunbound polypropylene back panel. Available in sizes SM to 4X. Elasticated wrists, waist and ankles. Stitched external seams in red. Thumb hole on sleeve. White.
  • The Tyvek™ front provides an ideal balance of protection, durability and comfort. It is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size. It is ultra-low-linting and antistatically treated. Silicon non-added.
  • The large back panel from shoulders to ankle is made of spunbound polypropylene: it is breathable and offers high level of comfort but no protection.
  • Tyvek™ 400 DualCombi is designed for work environment where comfort is important and exposure risk is limited to the front. Typical application is the handling of composite materials, paint spraying, glass manufacturing and utilities.
TRUSTED_SUSTAINABILITY
Specifica
Product Type Collared Coveralls
Size 2X-Large
Sesso Unisex
Materiale Polyethylene
Colore White
Monouso Single-use
Stile vita Elastic (Stitched-in)
Stile della caviglia Elastic
Stile polso Elastic
Stile collo Collared
Tipo di cappa Unhooded
Tipo di chiusura Front Zipper with Flap
Tipo di giuntura Stitched
Sterilità Non-sterile
Resistenza Liquid