Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Bag
GREENER_CHOICE
Codice prodotto. 17727887 Sfoglia Tutto Keystone Cleanroom Products Prodotti
Click to view available options
Dimensioni (L x P):
2 x 2 in.
3 x 1 in.
3 x 2 in.
3 x 2.5 in.
3 x 3 in.
4 x 1 in.
4 x 2 in.
5 x 1 in.
5 x 2 in.
5 x 3 in.
6 x 1 in.
6 x 2 in.
6 x 3 in.
7 x 1 in.
7 x 2 in.
8 x 1 in.
8 x 2 in.
8 x 3 in.
9 x 2 in.
9 x 3 in.
10 x 2 in.
10 x 3 in.
11 x 1 in.
11 x 2 in.
12 x 2 in.
12 x 3 in.
13 x 2 in.
Capacità (sistema imperiale):
3 oz.
4 oz.
5 oz.
6 oz.
7 oz.
7.5 oz.
8 oz.
9 oz.
10 oz.
11 oz.
14 oz.
15 oz.
16 oz.
18 oz.
20 oz.
22 oz.
24 oz.
26 oz.
27 oz.
30 oz.
36 oz.
Dimensione della confezione:
pezzi 1000
pezzi 250
pezzi 500
Codice prodotto. 17727887
Marca: Keystone Cleanroom Products BGT2A0312D
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. These filling needle bags stay in place via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifica
Autoclavabile Autoclavable
Capacità (sistema imperiale) 36 oz.
Certificazione/Conformità COC
Tipo di chiusura Drawstring
Colore White
Monouso Single-use
Da utilizzare con (applicazione) Steam Sterilization
Da utilizzare con (apparecchiatura) Pharmaceutical Equipment
Materiale Tyvek
Sterilità Non-sterile
Senza lattice Yes
Metodo di sterilizzazione Steam
Intervallo di temperatura (sistema imperiale) <250°F
Temperature Range (Metric) <125°C
Dimensioni (L x P) 12 x 3 in.
Lunghezza (sistema imperiale) 12 in.
Lunghezza (sistema metrico) 30.5 cm
Larghezza (sistema imperiale) 3 in.
Larghezza (sistema metrico) 7.6 cm
Product Type Autoclave Bag