Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Large Bag
GREENER_CHOICE
Codice prodotto. 17718047 Sfoglia Tutto Keystone Cleanroom Products Prodotti
Click to view available options
Dimensioni (L x P):
10 x 15 in.
10 x 34 in.
14 x 14 in.
16 x 16 in.
18 x 12 in.
18 x 26 in.
20 x 12 in.
20 x 16 in.
20 x 18 in.
20 x 20 in.
22 x 12 in.
22 x 14 in.
22 x 18 in.
24 x 10 in.
24 x 12 in.
24 x 14 in.
24 x 16 in.
24 x 18 in.
24 x 20 in.
24 x 24 in.
26 x 14 in.
28 x 12 in.
30 x 10 in.
30 x 12 in.
30 x 16 in.
30 x 18 in.
30 x 20 in.
32 x 12 in.
32 x 16 in.
32 x 18 in.
36 x 12 in.
36 x 14 in.
36 x 16 in.
38 x 14 in.
38 x 16 in.
40 x 14 in.
42 x 12 in.
64 x 10 in.
Dimensione della confezione:
pezzi 250
Codice prodotto. 17718047
Marca: Keystone Cleanroom Products BGT2A1820D
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Designed for steam sterilization of large or odd shaped pharmaceutical equipment change parts. Tyvek™ Autoclave Bag is closed via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically opened without having to cut or remove the particulate generating autoclave tape.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifica
Autoclavabile Autoclavable
Capacità (sistema imperiale) 340 oz.
Capacità (sistema metrico) 10 L
Certificazione/Conformità COC
Tipo di chiusura Drawstring
Colore White
Monouso Single-use
Da utilizzare con (applicazione) Steam Sterilization
Da utilizzare con (apparecchiatura) Pharmaceutical Equipment
Materiale Tyvek
Sterilità Non-sterile
Senza lattice Yes
Metodo di sterilizzazione Steam
Intervallo di temperatura (sistema imperiale) <250°F
Temperature Range (Metric) <125°C
Dimensioni (L x P) 20 x 18 in.
Lunghezza (sistema imperiale) 20 in.
Lunghezza (sistema metrico) 50.8 cm
Larghezza (sistema imperiale) 18 in.
Larghezza (sistema metrico) 45.7 cm
Product Type Autoclave Bag