Learn More
uvex U-Cap Sport Bump Cap - Long Brim
Sfoglia Tutto Uvex Prodotti
Click to view available options
Colore:
Black
Size:
52 to 54 cm
55 to 59 cm
60 to 63 cm
Dimensione della confezione:
Pezzo
Descrizione
- Ideal for areas where helmets are not compulsory, but there is still a risk of head injuries
- Innovative hard shell element in armadillo-design, honeycomb structure, and shock-absorbing elements
- Reliable impact protection in accordance with EN 812
- Long brim provides increased sun protection
Specifica
Specifica
| Colore | Black |
| Monouso | Reusable |
| Product Type | Bump Cap |
| Size | 52 to 54 cm |
| Include | Armadillo-Style Hard Shell |
| Ventilato | Vented |
| Head Style | Baseball Cap |