Learn More
uvex U-Fit Disposable NBR Protective Gloves
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
8
10
Dimensione della confezione:
Pezzo
Descrizione
- Thin, reliable NBR gloves enhanced grip capabilities due to the roughened palm surface
- Splash protection when handling chemicals such as acids, alkalis, solids, or aqueous salt solutions
- Silicone-free according to the impression test
- Highly flexible with good tactile sensitivity
- Very good mechanical strength
Specifica
Specifica
| Colore | Blue |
| Latex Free | Sì |
| Materiale | NBR |
| Palm Thickness (Metric) | 0.1 mm |
| Product Type | Chemical-resistant Gloves |
| Size | 8 |
| EN ISO 374-1 | Type B |
| EN ISO 374-1 CHEMICAL LETTER CODES | KPT |
| EN ISO 374 - 5 | Bacteria, Fungi WITH Virus Protection |