Learn More
Ub-K63 Antibody (HWA4C4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NBP2-00199-100ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Ub-K63 Monoclonal specifically detects Ub-K63 in Human samples. It is validated for Western Blot.
Specifica
| Ub-K63 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot < = 2 ug/ml | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains. | |
| Store at 4C. Do not freeze. |
| Western Blot | |
| HWA4C4 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| The immunogen for this antibody is Ub-K63. | |
| 100 μg | |
| Primary | |
| Human | |
| IgG2a |
Titolo del prodotto
For Research Use Only