missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Antibody (1B4-UB) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB600-776
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Ubiquitin Monoclonal specifically detects Ubiquitin in Human, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifica
| Ubiquitin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:1000, Simple Western, Flow Cytometry 1:10-1:50, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:500-1:1000 | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6233 | |
| Human, Bovine | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1B4-UB | |
| Unconjugated | |
| P0CG53 | |
| RPS27A | |
| Ubiquitin from bovine erythrocytes conjugated to bovine gamma globulin | |
| 0.05 mg | |
| Primary | |
| Reacts with Bovine. Cross reacts with Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto