Learn More
UCHL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12384-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.Specifica
| UCHL3 | |
| Polyclonal | |
| Unconjugated | |
| UCHL3 | |
| Ubiquitin carboxyl terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, UCHL3, UCH-L3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 498560, 50933, 7347 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P15374, Q91Y78, Q9JKB1 | |
| Uchl3 | |
| UCHL3 Fusion Protein Ag3053 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |