Learn More
Digi-Sense™ Ultra Low Freezer Verification Thermometer
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Temperature Range (Metric):
-90°C to 25°C
Dimensione della confezione:
Pezzo
Descripción
- Enclosed, unbreakable plastic bottle ensures accuracy
- Ideal for use in environmental chambers
- Individually serial number printed on the thermometer stem
- Non-mercury blue spirit liquid
- NIST-traceable statement of accuracy included
- Permanently fused markings
- Triangular-style, anti-roll fitting
- PFA-coated white-back glass
- Propylene glycol filled bottle
Especificaciones
Especificaciones
| Riempimento bottiglia | Propylene Glycol |
| Lunghezza (sistema metrico) complessiva | 145 mm |
| Riempimento termometro | Blue Spirit |
| Descrizione | PFA-coated, white-back glass ULT freezer varification thermometer |
| Temperature Range (Metric) | -90°C to 25°C |
| Da utilizzare con (apparecchiatura) | Freezers, refrigerators, incubators, ovens, environmental chambers or other environments where temperature stability is required |
| Accuracy | ±1.5°C; ±2.5°C below -50°C |
| Suddivisione | 1°C |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido