Learn More
OHAUS™ Ultra Multi-Purpose Clamps
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Dimensioni (sistema metrico) impugnatura:
0 to 103 mm
0 to 23 mm
0 to 32 mm
0 to 50 mm
0 to 70 mm
0 to 72 mm
Dimensione della confezione:
Pezzo
Descrizione
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Available as single or dual-adjust in nickel-plated zinc or stainless steel
- Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
Specifica
Specifica
| Tipo | Multi-purpose Clamp |
| Materiale | Nickel-plated |
| Include | Non-slip vinyl sleeves and fiberglass covers |
| Descrizione | Clamp, Multi Purpose, CLM-ULTRA3DZM |
| Diametro (sistema metrico) | 11 mm |
| Lunghezza (sistema imperiale) | 8.74 in. |
| Lunghezza (sistema imperiale) braccio | 5.000 in. |
| Lunghezza (sistema metrico) | 222 mm |
| Lunghezza (sistema metrico) braccio | 127 mm |
| Size | Medium |
| Vedi altri risultati |