Learn More
Fisherbrand™ Q125 Sonicator without Probe
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Modello:
Q125
Dimensione della confezione:
Pezzo
Descrizione
- Programmable operation to set time for hands-free operation
- Pulse mode to prevent heat build-up in temperature-sensitive samples
- Digital amplitude/intensity control from 20-100%
- Elapsed time indicator displays duration of sonication
- Display of wattage and joules for real-time energy monitoring
- Overload protection prevents damage to circuitry if a fault occurs
- Compact design to take up less space than competitive units
- Uses lead-free components
Specifica
Specifica
| Frequenza | 50/60 Hz |
| Tensione | 110 V |
| Include | Generator, converter, power cable, converter cable, wrench set |
| Modello | Q125 |
| Wattage | 125 w |
| Frequenza operativa | 20 kHz |