Learn More
Unc119b Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26201-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Mouse Unc119b is related closely to Unc119, a homolog of C. elegans unc119 protein which was first discovered in C. elegans on the basis of a spontaneous mutation affecting locomotion, feeding behavior and chemosensation. Mouse Unc119b shares 90% sequence identity with human UNC119b protein. UNC119b is a myristoyl-binding protein that acts as a cargo adapter: specifically binds the myristoyl moiety of a subset of N-terminally myristoylated proteins and is required for their localization. Myristoyl-binding activity of UNC119b is required to localize NPHP3 to the primary cilium.Specifica
| Unc119b | |
| Polyclonal | |
| Unconjugated | |
| UNC119B | |
| AA407659, AI414892, unc 119 homolog B (C. elegans), Unc119b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 106840, 611530, 84747 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A6NIH7, Q8C4B4 | |
| UNC119B | |
| Unc119b Fusion Protein Ag24599 | |
| 20 μL | |
| Primary | |
| Mouse, Canine, Human | |
| Antibody | |
| IgG |