Learn More
Abeba™ UNI6 1761 Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20345:2011 S2 SRC
- Functional leather with honeycomb pattern
- Black
- Steel toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350122)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- **Size 35 with replaceable acc Wave insole (art. 3576)
- Water-repellent
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20345:2011 S2 SRC |
| Colore | Black |
| Materiale | Leather |
| Size | 43 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 1761 |
| Materiale dello zoccolo | Steel |