Learn More
Abeba™ UNI6 36761 Shoes
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 O2 FO SRC
- Functional leather with honeycomb pattern
- Black
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Scratch-proof
- Water-repellent
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 O2 FO SRC |
| Colore | Black |
| Materiale | Leather |
| Size | 39 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 36761 |