Learn More
uvex Unigrip 6624 Cotton/Polyamide Assembly Gloves with PVC Dot Palm
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
7
8
9
Dimensione della confezione:
Pezzo
Descrizione
- Gloves are constructed of a 10 gauge cotton/polyamide blend
- PVC dots on the palm enhance grip capabilities in dry applications
- Ideal for assembly, sorting, and course mechanical work
Specifica
Specifica
| Coating Material | PVC Dots |
| Colore | Gray, Red |
| Monouso | Reusable |
| Finitura esterna | Smooth |
| Hand | Ambidestri |
| Materiale | Cotton, Polyamide |
| Powdered | Powder Free |
| Product Type | Work Gloves |
| Size | 7 |
| Sterilità | Non-sterile |
| Vedi altri risultati |