Learn More
Sampling Systems™ Unit Dose Sleeve Sampler Set
Codice prodotto. 15869261 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Materiale:
Stainless Steel (316L)
Dimensione della confezione:
Pezzo
Codice prodotto. 15869261
Marca: Sampling Systems™ 1500S01
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Developed for customers requiring accurate unit dose samples in small volumes. Sampling Systems™ Unit Dose Sleeve Sampler Set is ideal for use with a wide range of free flowing powders and granules.
  • For sampling small volume powders and granules
  • Chemically resistant 316L stainless steel construction
  • Different sample sizes can be taken using the same sampler
  • Includes the following:
    • 1 T bar Handle
    • 4 Handle Extension Rods
    • 1 Type 1 Sampler Chamber
    • 1 Type 2 Sample Chamber
    • 10 Sampling Tips
    • 1 High Quanity Sampler Case
  • All items included are available to be purchased individually
Specifica
Product Type Unit Dose Sampler Set
Materiale Stainless Steel (316L)
Include Handle, extension rod, sample chambers, sampling tips, and sampler case