Learn More
Corning™ Universal Fit Pipet Tips: Racked, Sterile
Sfoglia Tutto Corning Prodotti
Click to view available options
Volume (sistema metrico):
1 - 200 μl
1 to 200 μL
Da 100 a 1000 μl
Colore:
Blu
Incolore
Yellow
Dimensione della confezione:
pezzi 1000
pezzi 960
Descrizione
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifica
Specifica
| Volume (sistema metrico) | Da 100 a 1000 μl |
| Sterilità | Sì |
| Colore | Blu |
| Formato | In rack |
| Stile punta | Rotondo |
| Imballaggio | Rack |
| Graduato | Sì |
| Materiale | Polipropilene |