Learn More
Abnova™ UQCRH Recombinant Protein
Marca:  Abnova™ H00007388-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16131782
  • 528.00€ / 25µg
Spedizione stimata: 06-11-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifica
Specifica
AAH15177
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
35.8
8
Glutathione Sepharose 4 Fast Flow
25 ug
MGLEDEQKMLTESGDPEEEEEEEEELVDPLTTVREQCEQLEKCVKARERLELCDERVSSRSHTEEDCTEELFDFLHARDHCVAHKLFNNLK
MGC111572/QCR6
UQCRH
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
7388
UQCRH (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
UQCRH
Human
Recombinant
Solution