Learn More
UTP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18886-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Mediates apoptosis and actin stress fiber dissolution.Specifica
| UTP3 | |
| Polyclonal | |
| Unconjugated | |
| UTP3 | |
| CRL1, CRLZ1, Disrupter of silencing SAS10, SAS10, UTP3, UTP3 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305258, 57050, 65961 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AXX4, Q9JI13, Q9NQZ2 | |
| Utp3 | |
| UTP3 Fusion Protein Ag13454 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |