Learn More
uvex Unipur 6639 Polyurethane-Coated Assembly Gloves
Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
6
8
9
10
11
Dimensione della confezione:
10 paia
Pezzo
Description
- Polyamide gloves with polyurethane coating on the palm and fingertips
- Coating offers a high level of abrasion resistance
- Enhanced grip capabilities in dry, or slightly damp applications
- Lightweight and sensitive, ideal for precision mechanical work
- Dark colored material hides dirt and stains
Specifications
Specifications
| Coating Material | Polyurethane |
| Colore | Black |
| Finitura esterna | Smooth |
| Hand | Ambidestri |
| Materiale | Polyamide |
| Product Type | Mechanical Protection Gloves |
| Size | 6 |
| Stile | Full-hand Mitt |