Learn More
uvex Unipur 6639 Polyurethane-Coated Assembly Gloves
Codice prodotto. p-7160273 Sfoglia Tutto Uvex Prodotti
Click to view available options
Size:
6
8
9
10
11
Dimensione della confezione:
10 paia
Pezzo
Codice prodotto. 17220421
Marca: uvex 6024806
Unipur 6639 assembly gloves are suitable for precision mechanical work in dry to slightly damp work environments. The robust polyurethane coating provides abrasion-resistance, without compromising dexterity or tactility.
  • Polyamide gloves with polyurethane coating on the palm and fingertips
  • Coating offers a high level of abrasion resistance
  • Enhanced grip capabilities in dry, or slightly damp applications
  • Lightweight and sensitive, ideal for precision mechanical work
  • Dark colored material hides dirt and stains
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Colore Black
Finitura esterna Smooth
Hand Ambidestri
Materiale Polyamide
Product Type Mechanical Protection Gloves
Size 6
Stile Full-hand Mitt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.