Learn More
Greiner Bio-One VACUETTE™ K3EDTA Blood Collection Tubes
Click to view available options
Capacità (sistema metrico):
2 mL
3 mL
4 mL
6 mL
9 mL
Colore chiusura:
Lavender with Black Ring
Lavender with White Ring
Diametro (sistema metrico) esterno:
13 mm
16 mm
Tipo di chiusura:
Safety Pull Cap
Safety Twist Cap
Dimensione della confezione:
pezzi 100
pezzi 50
Descrizione
VACUETTE™ EDTA Blood Collection tubes are used for determinations in EDTA whole blood for haematology. Available with K2EDTA, K2EDTA Separator and K3EDTA. VACUETTE™ K3EDTA tubes contain tri-potassium K3EDTA which blocks the coagulation cascade.
- Tubes are supplied in packs of 50 pieces
- For venous blood collection
- Made out of virtually unbreakable PET plastic
- Improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
- Primarily in hematology
- Inner wall is coated with spray-dried K2EDTA which acts as an anticoagulant, binding calcium ions and interrupting the clotting cascade
- K2EDTA Blood Collection Tubes with Gel Separator and K3EDTA Blood Collection Tubes are available
- Pink caps for immunohematology are also available
- CE marked
Specifica
Specifica
| Product Type | Blood Collection Tube |
| Additivo | K3 EDTA |
| Materiale | PET |
| Sterilità | Sterile |
| Tipo di chiusura | Safety Pull Cap |
| Capacità (sistema metrico) | 2 mL |
| Da utilizzare con (apparecchiatura) | VACUETTE™ Blood Collection Needles |
| Colore chiusura | Lavender with White Ring |
| Altezza (sistema metrico) | 75 mm |
| Dimensioni provetta | 13 x 75 mm |
| Vedi altri risultati |