Learn More
BUCHI Vacuum Pump V-600 for Rotavapor R-220 Pro Industrial Evaporator for multiple benchtop Rotavapors
Sfoglia Tutto Buchi Prodotti
Click to view available options
Include:
Secondary Coil Condenser
Secondary Coil Condenser, Woulff Bottle
Secondary Cold Trap Condenser
Secondary Cold Trap Condenser, Woulff Bottle
Woulff Bottle
Dimensione della confezione:
Pezzo
Controllo:
I-300
I-300 Pro
Descrizione
- Chemically resistant 3-stage diaphragm pump
- Quiet and economical operation
- Optimally suited to work with the R-220 Pro, multiple Rotavapors or SyncorePlus Parallel Evaporation
- 1.5 mbar final vacuum
- 3.1 m3/hr. vacuum capacity
Specifica
Specifica
| Vuoto max | 1.5 mbar |
| Include | Secondary Coil Condenser, Woulff Bottle |
| Portata | 3.1 m³/hr. |
| Tipo di pompa | Oilless Diaphragm Pump |
| Tensione | 100/240 V |