Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Parallel
Codice prodotto. 10101811
Click to view available options
Foro spina:
2.5 mm
4 mm
6 mm
8 mm
10 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10101811
Marca: Lenz Laborglasintrumente 02942408
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12545 and in addition to DIN standard
  • With parallel side arms and hollow glass plug
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifica
Foro spina 8 mm
Materiale Borosilicate Glass (3.3)