missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAPB Mouse anti-Human, Clone: 4F6A6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66191-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.Specifica
| VAPB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95292 | |
| VAPB | |
| VAPB Fusion Protein Ag6313 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| 4F6A6 | |
| Unconjugated | |
| VAPB | |
| ALS8, VAMP associated protein B/C, VAMP B, VAMP B/VAMP C, VAMP C, VAP B, VAP B/VAP C, VAP C, VAPB | |
| Mouse | |
| Protein G | |
| RUO | |
| 9217 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto