Learn More
Kartell™ Variable Volume Pluripet Micropipettor
Ideal for dispensing large volumes of rather thick and viscous aqueous fluids. Kartell™ Variable Volume Pluripet Micropipettor is equipped with a special filter that protects the internal piston by increasing its lifetime and preventing contaminations.
369.00€ - 413.00€
Specifica
Certificazione/Conformità | EN IS 8655 |
---|---|
Autoclavabile | Autoclavable |
Tipo | Micropipette |
Codice del prodotto | Marca | Volume (sistema metrico) | Punte compatibili | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Volume (sistema metrico) | Punte compatibili | Prezzo | Quantità e disponibilità | ||||
10407912
|
Kartell™
0132910 |
1000 to 10,000 μL | Kartell™ 943 |
413.00€
Pezzo |
Spedizione stimata: 16-05-2024
Eseguire il login per visualizzare lo stock disponibile |
||||
10070782
|
Kartell™
0132210 |
100 to 1000 μL | Kartell™ 961, 941 |
369.00€
Pezzo |
Spedizione stimata: 24-06-2024
Eseguire il login per visualizzare lo stock disponibile |
||||
Descrizione
- The handle connection ensures a more comfortable and non-fatiguing grip
- The volume can be selected by turning the emptying button or turning the cylinder supplied for the purpose
- The large display ensures simple reading reading of the set volume value
- The button of each micropipettor has a specific colour so identification of the usable volume interval is simple
- The tip ejector is adjustable in height to house a large variety of tips and is can be disassembled to simplify cleaning operations
- The recalibration device ensures quick and precise recalibrations
- The robust structure of the PVDF body ensures high mechanical and chemical resistances
- Totally autoclavable at 121°C for 20 minutes
- The materials used for the structure are resistant and durable, and permit exposure to U.V. rays
Specifica
EN IS 8655 | |
Autoclavable | |
Micropipette |