Learn More
VACUUBRAND™ VARIO™ Chemistry Diaphragm Pump, MD 4C VARIO Select
GREENER_CHOICE
Codice prodotto. 16231782 Sfoglia Tutto Vacuubrand Prodotti
Click to view available options
Tipo di spina:
CEE
Dimensione della confezione:
Pezzo
Codice prodotto. 16231782
Marca: VACUUBRAND™ 20736550
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Provides pinpoint vacuum control for labs working with most common solvents by adjusting motor speed. The integrated VACUU·SELECT™ Controller provides an easy-to-use, application based interface that covers all common lab applications.
  • VACUU·SELECT Vacuum Controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Extraordinary diaphragm life for minimum operational and servicing costs
  • Meets high vacuum requirements for most high boiling solvents
  • Use manual set-point-control for simple processes, run fully automated distillations, or create unique applications with simple drag-and-drop editing
  • VACUU·SELECT Controller detects solvent boiling and automatically adjusts the pump's motor speed to maintain process control during solvent evaporation
  • VARIO Motor Speed Control provides exceptionally long maintenance intervals
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
TRUSTED_SUSTAINABILITY
Specifica
Vuoto max 1.5 Mbar (Without Gas Ballast), 3 Mbar (With Gas Ballast)
Controllo VACUU·SELECT
Peso di spedizione (sistema metrico) 24.1 kg
Include Ready-to-Use Pumping Unit (Completely Mounted with Manual)
Velocità motore 30-2400 min-1
Certificazione/Conformità CE-certification; II 3/-G IIC T3 x Internal Atm. Only
Dimensioni (L x P x A) 325 x 243 x 245 mm
Materiale PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM
Modello MD 4 C VARIO Select
Livello di rumorosità 43 dBA
Tipo di spina CEE
Wattage 0.53 kW
Peso (sistema metrico) 17.4 kg
Temperatura (sistema metrico) max 40°C
Temperatura min. (sistema metrico) 10°C
Portata 4.6 m³/hr.
Frequenza 50-60 Hz
Tipo di pompa Diaphragm Pump
Valori nominali armadio IP20
N. di teste pompe 4
Applicazioni Intelligent Vacuum with VARIO™-control, Rotary Evaporator/Parallel Evaporator, Vacuum Concentrators, Drying Chambers
No. of Stages 3
Maximum Pressure 1.1 bar
Tensione 200/230 V