Learn More
VACUUBRAND™ VARIO™ Chemistry Pumping Unit, PC 3012 VARIO Select
Codice prodotto. 16231802 Sfoglia Tutto Vacuubrand Prodotti
Click to view available options
Tipo di spina:
CEE
Kaltgerätestecker EN 60320
Modello:
PC 3012 VARIO Select
PC 3012 VARIO Select EKP
Dimensione della confezione:
Pezzo
Codice prodotto. 16231802
Marca: VACUUBRAND™ 20743850
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Provides pinpoint vacuum control by adjusting the motor speed. This model supports high pumping speeds and includes an exhaust vapor condenser. Suitable for use with floor-standing rotary evaporators, vacuum drying ovens, and work in miniplants, kilo labs, and scale-up facilities.
  • VACUU·SELECT Vacuum Controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Meets high vacuum requirements for large quantities of most high boiling solvents
  • Extraordinary diaphragm life for minimum operational and servicing costs
  • Excellent environmental compatibility through low energy consumption and efficient solvent recovery
  • High pumping speed and deep ultimate vacuum is suitable for demanding applications with high boiling point solvents
  • Use manual set-point-control for simple processes, run fully automated distillations, or create unique applications with simple drag-and-drop editing
  • For solvent evaporation, the VACUU·SELECT controller detects solvent boiling and automatically adjusts the pump's motor speed to maintain process control
  • VARIO motor speed control for exceptionally long maintenance intervals
  • Glass inlet separator with a robust protective coating prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
TRUSTED_SUSTAINABILITY
Specifica
Vuoto max 1.5 Mbar (Without Gas Ballast), 3 Mbar (With Gas Ballast)
Controllo VACUU·SELECT
Peso di spedizione (sistema metrico) 43.4 kg
Include Ready-to-Use Pumping Unit (Completely Mounted with Manual)
Velocità motore 30-2400 min-1
Certificazione/Conformità CE-certification; II 3/-G IIC T3 x Internal Atm. Only
Dimensioni (L x P x A) 616 x 387 x 450 mm
Descrizione Exhaust Vapor Condenser allows for near complete recovery of solvents when used with a chilled water line or recirculating chiller
Materiale PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM
Modello PC 3012 VARIO Select
Livello di rumorosità 47 dBA
Tipo di spina CEE
Wattage 1 kW
Temperatura (sistema metrico) max 40°C
Temperatura min. (sistema metrico) 10°C
Portata 15 m³/hr.
Frequenza 50-60 Hz
Tipo di pompa Diaphragm Pump
Valori nominali armadio IP40
N. di teste pompe 8
Applicazioni Intelligent Vacuum with VARIO™-control, Drying Chambers, Filtration and Solid Phase Extraction, Local Area Vacuum Networks for Laboratories
No. of Stages 3
Maximum Pressure 1.1 bar
Tensione 200/230 V