Learn More
Macherey-Nagel™ Supporto per precolonna VarioPrep
Codice prodotto. p-8012447 Sfoglia Tutto Macherey-Nagel Prodotti
Click to view available options
Diametro (sistema metrico) interno:
8 mm
32 mm
Lunghezza (sistema metrico):
10 mm
15 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 12757690
Marca: Macherey-Nagel™ 718253
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use VarioPrep™ stainless steel columns for HPLC. VarioPrep™ Guard Column Holder for Preperative HPLC come in different sizes for a variety of columns.
Features
  • Column system for preparative HPLC manufactured from stainless steel with two adjustable end fittings, e.g., for frequent use of backflushing techniques
  • Allows compensation of a dead volume, which could result at the column inlet after some time of operation, without need for opening the column
  • Can be packed with all NUCLEODUR™ and NUCLEOSIL™ spherical silicas
  • Column holders come in a variety of sizes for different columns
Specifica
Diametro (sistema metrico) interno 32 mm
Lunghezza (sistema metrico) 15 mm
Tipo Column Holder