Learn More
Eppendorf™ Varitip™ S Starter Kit
Sfoglia Tutto Eppendorf Prodotti
Click to view available options
:
Varitip S Starter Kit For Varipette 4720
Descrizione
- Eppendorf Varitip P is designed for aspirating 1 mL to 10 mL from beakers
- Eppendorf Varitip S for 2.5 mL to 10 mL forms a system with the Varitip and is used for aspirating liquid from tall, narrow-necked vessels
Specifica
Specifica
| Tipo | Eppendorf Varitips S Starter Kit |
| Da utilizzare con (applicazione) | Removing liquid from narrow-necked vessels and measuring flasks, |
| Include | 100 Maxitips, 10 dispensing parts, 10 valves |
| Quantità | 1 Kit |