missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VASN Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA545980
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Peptide sequence: TVTQLRPNAT YSVCVMPLGP GRVPEGEEAC GEAHTPPAVH SNHAPVTQAR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Rat: 92%.
The VASN gene encodes the protein vasorin, a type I transmembrane protein involved in cellular signaling, vascular development, and tissue repair. Vasorin is characterized by its expression in various tissues, including vascular endothelial cells and smooth muscle cells, where it functions to modulate cell proliferation and differentiation. It plays a significant role in the regulation of vascular remodeling and homeostasis by interacting with signaling molecules like transforming growth factor-beta (TGF-β), thereby influencing processes such as angiogenesis and wound healing. Vasorin is implicated in inhibiting TGF-β signaling pathways, which are crucial for maintaining vascular integrity and preventing excessive fibrosis during tissue repair. Dysregulation of VASN expression can contribute to pathological conditions, including cardiovascular diseases and cancer, where abnormal vascular remodeling and cell proliferation are evident. Research into vasorin continues to explore its functional mechanisms and potential therapeutic applications in diseases characterized by vascular dysregulation and tissue remodeling.
Specifica
| VASN | |
| Polyclonal | |
| Unconjugated | |
| VASN | |
| 2610528G05Rik; Atia; Protein slit-like 2; Slitl2; Slit-like 2; UNQ314/PRO357/PRO1282; VASN; Vasorin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114990 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6EMK4 | |
| VASN | |
| Synthetic peptide directed towards the C-terminal of human VASN (aa 521-570). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto