Learn More
Invitrogen™ VCP Monoclonal Antibody (5)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA3004
Descrizione
MA3-004 detects VCP protein in human, mouse, and rat samples. MA3-004 has successfully been used in Western blot immunoprecipitation, and immunohistochemical procedures. By Western blot, this antibody detects an ~97 kDa protein representing VCP from total lysate of cultured human B cells. The MA3-004 immunogen is a synthetic peptide corresponding to residues C G(792) G S V Y T E D N D D D L Y G(806) of mouse VCP. This peptide (Cat. # PEP-239) is available for use in neutralization and control experiments.
The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.
Specifica
| VCP | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| P46462, P55072, Q01853 | |
| VCP | |
| Synthetic peptide corresponding to residues C G(792) G S V Y T E D N D D D L Y G(806) of mouse VCP. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 5 | |
| Unconjugated | |
| VCP | |
| 15S Mg(2+)-ATPase p97 subunit; 3110001E05; AAA ATPase p97; ALS14; CDC48; cell division cycle gene CDC48; CMT2Y; epididymis luminal protein 220; epididymis secretory protein Li 70; HEL-220; HEL-S-70; homolog of yeast cdc48; IBMPFD; IBMPFD1; Inv protein; or HEL-S-70; p97; p97 atpase; p97/VCP; Protein CDC48; si:ch211-113n10.2; TER ATPase; TERA; transitional endoplasmic reticulum ATPase; valosin containing protein; valosin-containing protein; Vcp; wu:fd16d05; wu:fj63d11; yeast Cdc48p homolog | |
| Mouse | |
| Protein G | |
| RUO | |
| 116643, 269523, 7415 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.