Learn More
R&D Systems™ VersaClone cDNA, Human GlyT2/SLC6A5 (NP_004202)
SLC6A5 is found in glycinergic axons and maintains a high presynaptic pool of neurotransmitter at glycinergic synapses
Marca: R&D Systems™ RDC1231
Descrizione
The Human GlyT2/SLC6A5 (NP_004202) VersaClone cDNA clone contains the complete ORF for GlyT2/SLC6A5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GlyT2/SLC6A5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature
Specifica
| NP_004202 | |
| cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
| Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| 10 μg |
| 10 μg at 0.2 μg/μL | |
| 2406bp | |
| Human |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
For Reasearch Use Only; Not for Use in Humans