Learn More
Saint Gobain Life Sciences Tubo Versilic™
Codice prodotto. 10811081
Click to view available options
Diametro (sistema metrico) interno:
2 mm
3 mm
5 mm
10 mm
25 mm
Diametro (sistema metrico) esterno:
5.5 mm
6 mm
7 mm
9 mm
18 mm
35 mm
Spessore (sistema metrico):
1.25 mm
2 mm
4 mm
5 mm
Overall Length (Metric):
10 m
25 m
50 m
Dimensione della confezione:
10m
25m
50m
Pezzo
Codice prodotto. 10811081
Marca: Saint Gobain Life Sciences 760250
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use this tubing for biopharmaceutical applications. Versilic™ Silicone Tubing is biologically inert and does not inhibit cell culture. Stays flexible down to -50°C and maintains its chemical, mechanical and electrical performance up to +200°C.
  • Meets the European Pharmacopoeia requirements for silicone elastomer for closures and tubing, USP Class VI biological tests and ISO 10993 biocompatibility (systemic toxicity, irritation, sensitization, citotoxicity, hemocompatibility and Ames test)
  • Meets FDA and BfR positive lists as well as food grade tests according to EC directives
  • Peroxide-cured silicone
  • Biological neutrality
  • FDA/BfR
  • US and European Pharmacopoeia
  • Temperature resistance -50 to +200°C
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità EU Pharmacopoeia, USP Class VI, ISO 10993
Diametro (sistema imperiale) interno 0.12 in.
Diametro (sistema metrico) interno 3 mm
Materiale Versilic
Diametro (sistema imperiale) esterno 0.28 in.
Diametro (sistema metrico) esterno 7 mm
Overall Length (English) 164 ft.
Overall Length (Metric) 50 m
Spessore (sistema imperiale) 0.08 in.
Spessore (sistema metrico) 2 mm
Min. Operating Temperature (Metric) °
Quantità confezione 1 Ea.
Durezza 60 (Shore A)
Max. Operating Temperature (Metric) 200°C
Product Type Tubing