Learn More
Tocris Bioscience™ VH 032 amide-alkylC6-acid
Click to view available options
Quantità:
25 mg
Dimensione della confezione:
25mg
Descrizione
VH 032 amide-alkylC6-acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC™ research and development; incorporates an E3 ligase ligand plus alkylC6 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifica
Specifica
| Materiale o nome chimico | 8-((-1-((2 S,4 R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-8-oxooctanoic acid |
| CAS | 2172819-75-7 |
| Quantità | 25 mg |
| Bersaglio | Degrader Building Blocks |
| Formula molecolare | C30H42N4O6S |
| Purezza | 0.95 |