Learn More
Tocris Bioscience™ VH 032, thiol
Click to view available options
Quantità:
50 mg
Dimensione della confezione:
50mg
Descrizione
VH 032, thiol is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with thiol group for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifica
Specifica
| Materiale o nome chimico | (2S,4R)-1-((R)-2-Acetamido-3-mercapto-3-methylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2098836-54-3 |
| Quantità | 50 mg |
| Bersaglio | Degrader Building Blocks |
| Formula molecolare | C23H30N4O4S2 |
| Purezza | 0.95 |