Learn More
Buerkle™ Rapid Couplings, NW 3.2 mm, Male Connections, Hose Nipple with Hose Screw-Joint
Codice prodotto. 10645147 Sfoglia Tutto Bürkle Prodotti
Click to view available options
Dimensioni dei tubi:
2 mm dia. x 27 mm
4 mm dia. x 37 mm
Materiale:
POM
Polypropylene
Descrizione:
With valve
Without valve
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10645147
Marca: Buerkle™ 87521142
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Rapid Couplings, NW 3.2 mm, Male Connections, Hose Nipple with Hose Screw-Joint – Fast and safe connection for liquids, gases, compressed air and vacuum. Buerkle™ Rapid Couplings are ideal when connecting and separating hoses, machines, apparatus, containers and more.
  • Manufactured from polyoxymethylene or polypropylene
  • Maximum flow possible
  • Easy single-handed operation
  • Secure O-ring guarantees that couplings are leakproof
  • Polyoxymethylene operating pressure at 20°C: up to 10 bar, operating temperature from −40° to +80°C
  • Polypropylene operating pressure at 20°C: up to 8 bar, operating temperature from 0° to 110°C
  • Couplings simply connect to each other, automatically clicking into place and opening the valves
  • When disconnecting, the valves are automatically sealed, no leakage occurs
TRUSTED_SUSTAINABILITY
Specifica
Descrizione With valve
Diametro (sistema metrico) interno 2 mm
Materiale POM
Diametro (sistema metrico) esterno 4 mm
Overall Length (Metric) 27 mm
Da utilizzare con 2 x 4 mm Hose
Min. Operating Temperature (Metric) °
Quantità confezione 1
Dimensioni dei tubi 2 mm dia. x 27 mm
Larghezza (sistema metrico) 3.2 mm
Include Rapid coupling
Pressione massima 10 bar
Max. Operating Temperature (Metric) 80°C
Product Type Rapid Coupling