Learn More
1-Amino-4-hydroxyanthraquinone, 96%, Thermo Scientific™
Marca:  Thermo Scientific Alfa Aesar B24388.22
Quantità 100g
Codice prodotto. 11455827
  • ONLINE EXCLUSIVE PRICE
    291.00€
    288.99€ / 100g
    Risparmia 2.01€
    0.69% Sconto
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.
Solubility
Soluble in water, ethanol, acetone, benzene.
Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
1-Amino-4-hydroxyanthraquinone
C14H9NO3
100g
Soluble in water,ethanol,acetone,benzene.
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N
239.23
239.23
116-85-8
MFCD00001223
1-amino-4-hydroxyanthraquinone, disperse red 15, calcosyn pink b, celutate pink b, nacelan pink b, amacel pink b, celutate pink bn, celutate pink by, cibacete red 3b, disperse red 2s
AQXYVFBSOOBBQV-UHFFFAOYSA-N
1-amino-4-hydroxyanthracene-9,10-dione
8323
96%
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS Documenti