Learn More
Invitrogen™ CGBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA530606
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: HepG2. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (84%), Rhesus Monkey (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Proteins that contain a CXXC motif within their DNA-binding domain, such as CXXC1, recognize CpG sequences and regulate gene expression (Carlone and Skalnik, 2001).
Specifikationer
| CGBP | |
| Polyclonal | |
| Unconjugated | |
| CXXC1 | |
| 2410002I16Rik; 5830420C16Rik; AI426635; CFP 1; CFP1; CGBP; CpG binding protein; cpG-binding protein; cpG-binding protein; CXXC-type zinc finger protein 1; CXXC 1; CXXC finger 1; CXXC finger 1 (PHD domain); CXXC finger protein 1; Cxxc1; CXXC-type zinc finger protein 1; DNA-binding protein with PHD finger and CXXC domain; hCGBP; HsT2645; PCCX 1; Pccx1; PHD finger and CXXC domain-containing protein 1; PHF 18; PHF18; SPP 1; SPP1; ZCGPC 1; ZCGPC1; zinc finger, CpG binding-type containing 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30827 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.74 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9P0U4 | |
| CXXC1 | |
| Recombinant protein fragment corresponding to a region within amino acids 283 and 522 of Human CGBP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering