Learn More
JUN (phospho T239), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of JUN.
Marca: Abnova PAB12615.100ug
Descrizione
This gene is the putative transforming gene of avian sarcoma virus 17. It encodes a protein which is highly similar to the viral protein, and which interacts directly with specific target DNA sequences to regulate gene expression. This gene is intronless and is mapped to 1p32-p31, a chromosomal region involved in both translocations and deletions in human malignancies. [provided by RefSeq
Sequence: GETPPSpecifica
| JUN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of JUN. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| AP-1/AP1/c-Jun | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼48 KDa of human JUN at a phosphorylation site of Theronine 239. It does not react to non-phosphorylated peptides. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| JUN | |
| JUN | |
| Synthetic phosphopeptide corresponding to residues surrounding T239 of human JUN. | |
| RUO | |
| 3725 | |
| Human, Mouse, Rat | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only