Learn More
Ansell MICRO-TOUCH™ Sterile Gloves
Codice prodotto. 17863622
Click to view available options
Size:
6.5/7
7.5/8
8.5/9
Dimensione della confezione:
50 paia
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
This product contains natural rubber latex which may cause allergic reactions, including anaphylactic responses.
Codice prodotto. 17863622
Marca: Ansell 553342
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
This product contains natural rubber latex which may cause allergic reactions, including anaphylactic responses.
Latex, powder-free gloves with high comfort and grip for use in industrial applications.
  • An inner polyacrylate coating creates a softer feel on the user's hands, making donning easier even with wet hands
  • Natural rubber latex formulation ensures a comfortable, well-fitting glove
  • Extensive leaching phases ensure a very low residual protein content
  • Textured fingers provide a safe and secure grip in wet or dry conditions
  • High tactile sensitivity and dexterity
  • Packaged in pairs with a protective double wrappolyisopreneng
  • Minimizes Type I allergy
  • For use in industrial applications
Specifica
Certificazione/Conformità CE 2797, ISO 13485, ISO 9001, ISO 10993-10:2004
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 455 1-4
Accelerator Free No
Applicazioni Industrial
AQL 1.5
Colore White
Monouso Single-use
Finitura esterna Fully Textured
Finger Thickness (English) 6.7 mil
Finger Thickness (Metric) 0.17 mm
Latex Free No
Materiale Latex
Palm Thickness (English) 5.9 mil
Palm Thickness (Metric) 0.15 mm
Powdered Powder Free
Product Type Latex Gloves
Size 6.5/7
Sterilità Sterile
Spessore (sistema imperiale) 5.9 mil
Spessore (sistema metrico) 0.15 mm
Modello MICRO-TOUCH-323039
Lunghezza (sistema imperiale) 9.6 in.
Lunghezza (sistema metrico) 245 mm