Learn More
Tocris Bioscience™ OUL 35
Click to view available options
Quantità:
5 mg
25 mg
Dimensione della confezione:
25mg
5mg
Descrizione
OUL 35 is a selective PARP-10 (also known as ADP-ribosyltransferase Diptheria Toxin-like 10, ARTD10) inhibitor (IC50 = 0.329 μM). Displays selectivity for PARP-10 over a panel of PARP family members. Rescues HeLa cells from PARP-10-induced cell death. Sensitizes cancer cells to hydroxyurea-induced genotoxic stress.
Specifica
Specifica
| CAS | 6336-34-1 |
| Sinonimo | 4,4′-Oxybis[benzamide] |
| Quantità | 5 mg |
| Formula molecolare | C14H12N2O3 |
| Peso formulazione | 256.26 |
| Purezza | 98% |