Learn More
RNA binding motif protein, X-linked 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-83453-0.1ML
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
RNA binding motif protein, X-linked 2 Polyclonal specifically detects RNA binding motif protein, X-linked 2 in Human samples. It is validated for Western Blot.
Specifica
| RNA binding motif protein, X-linked 2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51634 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| CGI-79,2810411A03Rik, RNA binding motif protein, X-linked 2, RNA-binding motif protein, X-linked 2 | |
| The immunogen is a synthetic peptide directed towards the middle region of human RNA binding motif protein, X-linked 2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |