Learn More
Novus Biologicals™ Recombinant Human Band 3 GST (N-Term) Protein
Codice prodotto. 18651251 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
10 μg
25 μg
Dimensione della confezione:
10µg
25µg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18651251
Marca: Novus Biologicals™ H00006521Q0125ug
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Recombinant Protein
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (applicazione) Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification
Formulazione 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
ID gene (immissione) 6521
Peso molecolare MolecularWeight-theroretical: 36.74 kDa
Quantità 25 μg
Requisiti di stoccaggio Store at -80°C. Avoid freeze-thaw cycles.
Alias gene AE 1, AE1MGC126619, Anion exchange protein 1, Anion exchanger 1, band 3 anion transport protein, BND3, CD233, CD233 antigen, DI, EMPB3, EPB3MGC126623, erythrocyte membrane protein band 3, erythroid anion exchange protein, FR, Froese blood group, MGC116750, MGC116753, RTA1A, Solute carrier family 4 member 1, solute carrier family 4, anion exchanger, member 1 (erythrocyte membraneprotein band 3, Diego blood group), solute carrier family 4, anion exchanger, number 1, SW, Swann blood group, Waldner blood group, WD, WD1, WR, Wright blood group
Simbolo del gene SLC4A1
Research Category Phospho Specific
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Proteine Band 3