Learn More
Vitronectin Antibody (102), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP2-89488APC
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Vitronectin Monoclonal antibody specifically detects Vitronectin in Human samples. It is validated for ELISA
Specifica
| Vitronectin | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 102 | |
| ELISA | |
| complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Vitronectin (Accession#: NP_000629.3; Asp20-Leu478). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 7448 | |
| Store at 4C in the dark. | |
| IgG |