Learn More
ABCA7 Antibody (7A1-144), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NB400-163JF646
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ABCA7 Monoclonal antibody specifically detects ABCA7 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifica
| ABCA7 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 7A1-144 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ABCA-SSN, ABCXATP-binding cassette sub-family A member 7, ATP-binding cassette, sub-family A (ABC1), member 7, Autoantigen SS-N, EC 2.2.1.1, EC 3.6.3, EC 3.6.3.41, FLJ40025, Macrophage ABC transporter | |
| Mouse ABCA7 stably-transfected HeLa cells. [UniProt# Q91V24] | |
| 0.1 mL | |
| ABC Transporters, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 10347 | |
| Store at 4C in the dark. | |
| IgG2a κ |